Gene Transcripts

Gene Symbol
TMEM268
Entrez Gene ID
203197
Gene Name
transmembrane protein 268
Gene Aliases
C9orf91
Chromosome Location
Chr. 9: 114603897 - 114646423 on Build GRCh38
UniGene ID
Hs.522357
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153045.3 NP_694590.2 9 3471
XM_006716997.1 XP_006717060.1 9 3474
XM_011518350.1 XP_011516652.1 10 3642
XM_011518351.2 XP_011516653.1 10 3770
XM_011518352.1 XP_011516654.1 10 3639
XM_011518353.1 XP_011516655.1 9 3531
XM_011518354.1 XP_011516656.1 9 3677
XM_011518355.1 XP_011516657.1 9 3674
XM_017014429.1 XP_016869918.1 10 3798
XM_017014430.1 XP_016869919.1 8 3563
GenBank mRNA AF038174.1 - 1 331
AK098022.1 - 1 2005
BC019913.1 - 3 2643

View Full Product Details