Gene Transcripts

Gene Symbol
Sprr2i
Entrez Gene ID
20763
Gene Name
small proline-rich protein 2I
Gene Aliases
Chromosome Location
Chr. 3: 92407991 - 92409271 on Build GRCm38
UniGene ID
Mm.196439
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011475.3 NP_035605.1 2 464
GenBank mRNA AY158993.1 - 2 464

View Full Product Details