Gene Transcripts

Gene Symbol
Rab12
Entrez Gene ID
19328
Gene Name
RAB12, member RAS oncogene family
Gene Aliases
2900054P15Rik, AW555448, C77700
Chromosome Location
Chr. 17: 66494512 - 66519934 on Build GRCm38
UniGene ID
Mm.248313
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024448.2 NP_077768.2 6 750
GenBank mRNA AB232607.1 - 6 650
AK013690.1 - 5 406

View Full Product Details