Gene Transcripts
- Gene Symbol
- Mybl2
- Entrez Gene ID
- 17865
- Gene Name
- myeloblastosis oncogene-like 2
- Gene Aliases
- B-Myb, Bmyb
- Chromosome Location
- Chr. 2: 163054333 - 163084688 on Build GRCm38
- UniGene ID
- Mm.4594
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008652.2 |
NP_032678.1
|
14 |
2869 |
|
GenBank mRNA |
AK028497.1 |
- |
14 |
2869 |
|
View Full Product Details