Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026171.1 | NP_080447.1 | 6 | 401 | |
GenBank mRNA | AK004676.1 | - | 6 | 433 | |
AK013982.1 | - | 6 | 400 | ||
AK018049.1 | - | 6 | 397 | ||
AK140509.1 | - | 6 | 432 | ||
BC031847.1 | - | 6 | 384 | 3964470 |