Gene Transcripts

Gene Symbol
NHLH1
Entrez Gene ID
4807
Gene Name
nescient helix-loop-helix 1
Gene Aliases
HEN1, NSCL, NSCL1, bHLHa35
Chromosome Location
Chr. 1: 160367071 - 160372848 on Build GRCh38
UniGene ID
Hs.30956
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005598.3 NP_005589.1 2 1780
GenBank mRNA BC013789.1 - 2 1767
M96739.1 - 2 1783

View Full Product Details