Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177178.3 | NP_796152.2 | 4 | 526 | |
GenBank mRNA | AK036527.1 | - | 4 | 547 | |
AK079450.1 | - | 4 | 526 | ||
AK153728.1 | - | 4 | 477 | ||
BC095971.1 | - | 4 | 443 | 30108856 | |
BC120727.1 | - | 4 | 498 | 40129650 | |
BC125421.1 | - | 4 | 498 | 40129936 |