Gene Transcripts
- Gene Symbol
- A530021J07Rik
- Entrez Gene ID
- 330578
- Gene Name
- Riken cDNA A530021J07 gene
- Gene Aliases
- Chromosome Location
- Chr. 7: 83148463 - 83179432 on Build GRCm38
- UniGene ID
- Mm.5796
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK040731.1 |
- |
5 |
927 |
|
View Full Product Details