Gene Transcripts

Gene Symbol
Slc26a7
Entrez Gene ID
208890
Gene Name
solute carrier family 26, member 7
Gene Aliases
Chromosome Location
Chr. 4: 14502430 - 14621971 on Build GRCm38
UniGene ID
Mm.483839
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145947.2 NP_666059.2 18 1984
GenBank mRNA AF345194.1 - 17 1886
AK054327.1 - 18 1992
BC026928.1 - 18 1963 4020760

View Full Product Details