Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172778.2 | NP_766366.2 | 9 | 1059 | |
GenBank mRNA | AK031833.1 | - | 9 | 1043 | |
AK054050.1 | - | 9 | 1058 | ||
BC113182.1 | - | 9 | 1048 | 40061991 | |
BC113788.1 | - | 9 | 1048 | 40061990 |