Gene Transcripts

Gene Symbol
Ffar4
Entrez Gene ID
107221
Gene Name
free fatty acid receptor 4
Gene Aliases
AI552415, GT01, Gpr120, Gpr129, KPG_013, O3far1, Pgr4
Chromosome Location
Chr. 19: 38097079 - 38114263 on Build GRCm38
UniGene ID
Mm.35288
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181748.2 NP_861413.1 3 1183
GenBank mRNA AB115769.1 - 3 1135
AK131616.1 - 3 1170
BC053698.1 - 3 1183 6529120

View Full Product Details