Gene Transcripts

Gene Symbol
HTR1E
Entrez Gene ID
3354
Gene Name
5-hydroxytryptamine receptor 1E
Gene Aliases
5-HT1E
Chromosome Location
Chr. 6: 86936919 - 87020068 on Build GRCh38
UniGene ID
Hs.1611
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000865.2 NP_000856.1 2 1827
XM_011535789.2 XP_011534091.1 2 2275
GenBank mRNA BC069751.1 - 2 1406
M91467.1 - 3 1690

View Full Product Details