Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023630.2 | NP_076119.2 | 2, 3 | 182 | |
GenBank mRNA | AF250835.1 | - | 2, 3 | 150 | |
AK005881.1 | - | 2, 3 | 157 | ||
BC050756.1 | - | 2, 3 | 165 | 6741860 | |
BY092414.1 | - | 2, 3 | 182 |