Gene Transcripts

Gene Symbol
Usp50
Entrez Gene ID
75083
Gene Name
ubiquitin specific peptidase 50
Gene Aliases
1700086G18Rik, 4930511O11Rik
Chromosome Location
Chr. 2: 126761050 - 126783460 on Build GRCm38
UniGene ID
Mm.389373
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029163.3 NP_083439.2 5 882
GenBank mRNA AK015766.1 - 5 867
BC061020.1 - 5 859 6772471

View Full Product Details