Gene Transcripts

Gene Symbol
Rpl37
Entrez Gene ID
67281
Gene Name
ribosomal protein L37
Gene Aliases
3110005M08Rik
Chromosome Location
Chr. 15: 5116613 - 5119140 on Build GRCm38
UniGene ID
Mm.10474
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026069.3 NP_080345.1 4 729
GenBank mRNA AK011101.1 - 4 690
AK014003.1 - 4 698

View Full Product Details