Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_197982.3 | NP_932099.2 | 3 | 485 | |
GenBank mRNA | AK011285.1 | - | 2 | 325 | |
AK012002.1 | - | 3 | 395 | ||
AK016960.1 | - | 3 | 395 | ||
AK137114.1 | - | 391 | |||
AK145927.1 | - | 3 | 428 | ||
AK146294.1 | - | 3 | 430 | ||
AK149475.1 | - | 3 | 391 | ||
AK150997.1 | - | 3 | 414 | ||
AK151395.1 | - | 3 | 395 | ||
AK152394.1 | - | 3 | 395 | ||
AK162752.1 | - | 953 | |||
AK164289.1 | - | 3 | 355 | ||
AK167890.1 | - | 3 | 355 | ||
AK168079.1 | - | 3 | 354 | ||
AK172448.1 | - | 3 | 414 | ||
BC020134.1 | - | 3 | 373 | 3969048 | |
CF172497.1 | - | 3 | 382 |