Gene Transcripts
- Gene Symbol
- Timm50
- Entrez Gene ID
- 66525
- Gene Name
- translocase of inner mitochondrial membrane 50
- Gene Aliases
- 2810403L02Rik, AU015082, TIM50L
- Chromosome Location
- Chr. 7: 28305516 - 28312046 on Build GRCm38
- UniGene ID
- Mm.167913
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025616.3 |
NP_079892.1
|
4, 5 |
333 |
|
GenBank mRNA |
AK008340.1 |
- |
4, 5 |
334 |
|
AK008382.1 |
- |
4, 5 |
331 |
|
AK010832.1 |
- |
4, 5 |
299 |
|
AK012977.1 |
- |
4, 5 |
330 |
|
AK088074.1 |
- |
4, 5 |
326 |
|
AK167473.1 |
- |
4, 5 |
336 |
|
AK178698.1 |
- |
3, 4 |
107 |
|
AY551342.1 |
- |
4, 5 |
319 |
|
BC010303.1 |
- |
4, 5 |
308 |
3597676 |
BC037056.1 |
- |
4, 5 |
335 |
|
View Full Product Details