Gene Transcripts

Gene Symbol
Mtmr7
Entrez Gene ID
54384
Gene Name
myotubularin related protein 7
Gene Aliases
Chromosome Location
Chr. 8: 40550159 - 40634792 on Build GRCm38
UniGene ID
Mm.294871
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040699.1 NP_001035789.1 6, 7 804
GenBank mRNA AK028160.1 - 6, 7 770
AK081973.1 - 6, 7 790
AK148768.1 - 6, 7 805
BC032254.1 - 6, 7 806 5368816

View Full Product Details