Gene Transcripts

Gene Symbol
Rab9
Entrez Gene ID
56382
Gene Name
RAB9, member RAS oncogene family
Gene Aliases
2410064E05Rik, AI195561, Rab9a, Sid6061p
Chromosome Location
Chr. X: 166457251 - 166479867 on Build GRCm38
UniGene ID
Mm.25306
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019773.2 NP_062747.1 3 823
GenBank mRNA AB027290.1 - 3 652
AB232602.1 - 1 437
AK010710.1 - 3 602
AK017301.1 - 3 617
AK032133.1 - 3 588
BC008160.1 - 3 622 3964134

View Full Product Details