Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190161.1 | NP_001177090.1 | 8 | 1157 | |
NM_019976.3 | NP_064360.1 | 8 | 1154 | ||
GenBank mRNA | AF156598.1 | - | 8 | 1149 | |
AK012815.1 | - | 3 | 264 | ||
AK012895.1 | - | 3 | 261 | ||
AK017302.1 | - | 8 | 1096 | ||
AK041835.1 | - | 8 | 1104 | ||
AK167415.1 | - | 8 | 1100 | ||
BC019165.1 | - | 8 | 1112 | 3963608 |