Gene Transcripts

Gene Symbol
Zfp330
Entrez Gene ID
30932
Gene Name
zinc finger protein 330
Gene Aliases
BC008086, Noa36, Znf330
Chromosome Location
Chr. 8: 82763620 - 82774126 on Build GRCm38
UniGene ID
Mm.269248
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145600.1 NP_663575.1 5 345
GenBank mRNA AK029489.1 - 4 170
AK037748.1 - 1 627
AK050534.1 - 5 355
AK086617.1 - 5 379
BC008086.1 - 5 345 3586161
BC030675.1 - 5 343 1068780
BC037025.1 - 1 1702
BC083081.1 - 5 379 6510396

View Full Product Details