Gene Transcripts
- Gene Symbol
- Psg17
- Entrez Gene ID
- 26437
- Gene Name
- pregnancy specific glycoprotein 17
- Gene Aliases
- Cea-2, Cea2, mmCGM5
- Chromosome Location
- Chr. 7: 18813937 - 18821591 on Build GRCm38
- UniGene ID
- Mm.302844
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M83344.1 |
- |
5 |
1358 |
|
View Full Product Details