Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009541.2 | NP_033567.2 | 3 | 410 | |
GenBank mRNA | AK051971.1 | - | 3 | 400 | |
AK134251.1 | - | 3 | 370 | ||
BC060055.1 | - | 3 | 384 | ||
BC145312.1 | - | 2 | 283 | 9053867 | |
BC145313.1 | - | 2 | 283 | ||
BC145821.1 | - | 2 | 283 | 40131124 | |
U14556.1 | - | 3 | 408 | ||
U22396.1 | - | 3 | 398 |