Gene Transcripts
- Gene Symbol
- LILRB5
- Entrez Gene ID
- 10990
- Gene Name
- leukocyte immunoglobulin like receptor B5
- Gene Aliases
- CD85C, LIR-8, LIR8
- Chromosome Location
- Chr. 19: 54250405 - 54257308 on Build GRCh38
- UniGene ID
- Hs.655338
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001081442.2 |
NP_001074911.1
|
11, 12 |
1686 |
|
NM_001081443.2 |
NP_001074912.1
|
10, 11 |
1386 |
|
NM_001304457.1 |
NP_001291386.1
|
11, 12 |
1659 |
|
NM_006840.4 |
NP_006831.1
|
11, 12 |
1683 |
|
XM_011526359.2 |
XP_011524661.1
|
11, 12 |
1670 |
|
XM_011526360.2 |
XP_011524662.1
|
11, 12 |
1667 |
|
XM_011526361.2 |
XP_011524663.1
|
10, 11 |
1619 |
|
XM_011526362.2 |
XP_011524664.1
|
10, 11 |
1351 |
|
GenBank mRNA |
AF025534.1 |
- |
11, 12 |
1634 |
|
AK057072.1 |
- |
10, 11 |
1340 |
|
AK092867.1 |
- |
10, 11 |
2584 |
|
AK096122.1 |
- |
7, 8 |
2059 |
|
AK223296.1 |
- |
11, 12 |
1642 |
|
AK290845.1 |
- |
11, 12 |
1667 |
|
AK301112.1 |
- |
11, 12 |
1622 |
|
BC025704.1 |
- |
11, 12 |
1637 |
5223581 |
View Full Product Details