Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286416.1 | NP_001273345.1 | 3 | 447 | |
NM_006936.2 | NP_008867.2 | 3 | 333 | ||
GenBank mRNA | AK222591.1 | - | 3 | 264 | |
AK300822.1 | - | 3 | 399 | ||
AK312350.1 | - | 3 | 264 | ||
BC000036.2 | - | 3 | 258 | 3505840 | |
BC008420.1 | - | 3 | 333 | 4091864 | |
BT007008.1 | - | 3 | 172 | ||
CR542173.1 | - | 3 | 172 | ||
CR542188.1 | - | 3 | 172 | ||
X99584.1 | - | 3 | 266 |