Gene Transcripts

Gene Symbol
Slc36a2
Entrez Gene ID
246235
Gene Name
solute carrier family 36 (proton/amino acid symporter), member 2
Gene Aliases
Pat2, Tramd1
Chromosome Location
Chr. 10: 40497184 - 40525033 on Build Rnor_6.0
UniGene ID
Rn.145405
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139339.1 NP_647555.1 1, 2 262
GenBank mRNA AF512430.1 - 1, 2 262
BC166414.1 - 1, 2 196 9027758

View Full Product Details