Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_053634.1 | NP_446086.1 | 6, 7 | 480 | |
GenBank mRNA | AB036792.1 | - | 6, 7 | 480 | |
BC088123.1 | - | 6, 7 | 472 | 7376834 | |
FQ220557.1 | - | 6, 7 | 483 | ||
FQ233041.1 | - | 6, 7 | 483 |