Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_199102.1 | NP_954533.1 | 5 | 647 | |
GenBank mRNA | BC061783.1 | - | 5 | 647 | |
FQ214531.1 | - | 5 | 651 | ||
FQ219988.1 | - | 5 | 660 | ||
FQ220693.1 | - | 5 | 645 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_002730236.3 | XP_002730282.1 | 1 | 624 |