Gene Transcripts

Gene Symbol
Cnppd1
Entrez Gene ID
316530
Gene Name
cyclin Pas1/PHO80 domain containing 1
Gene Aliases
RGD735175
Chromosome Location
Chr. 9: 82338861 - 82345262 on Build Rnor_6.0
UniGene ID
Rn.101111
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199112.1 NP_954543.1 9 1303
XM_006245226.2 XP_006245288.1 9 1337
GenBank mRNA BC061750.1 - 9 1303

View Full Product Details