Gene Transcripts

Gene Symbol
RXFP1
Entrez Gene ID
59350
Gene Name
relaxin/insulin like family peptide receptor 1
Gene Aliases
LGR7, RXFPR1
Chromosome Location
Chr. 4: 158521711 - 158653372 on Build GRCh38
UniGene ID
Hs.591686
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253727.1 NP_001240656.1 14 1434
NM_001253728.1 NP_001240657.1 13 1254
NM_001253729.1 NP_001240658.1 12 1209
NM_001253730.1 NP_001240659.1 13 1275
NM_001253732.1 NP_001240661.1 15 1437
NM_001253733.1 NP_001240662.1 14 1365
NM_021634.3 NP_067647.2 14 1353
NR_045579.1 14 1970
NR_045580.1 14 1406
NR_045581.1 14 1377
NR_045582.1 12 1314
NR_045583.1 13 1293
NR_045584.1 15 1406
XM_011532174.1 XP_011530476.1 14 1400
XM_011532175.1 XP_011530477.1 13 1331
XM_011532176.1 XP_011530478.1 13 1253
XM_011532179.2 XP_011530481.1 14 1403
XM_017008517.1 XP_016864006.1 13 1328
XM_017008518.1 XP_016864007.1 14 1322
XM_017008519.1 XP_016864008.1 15 1367
XM_017008520.1 XP_016864009.1 14 1456
XM_017008521.1 XP_016864010.1 15 1286
XM_017008522.1 XP_016864011.1 14 1371
XM_017008523.1 XP_016864012.1 14 1325
XM_017008524.1 XP_016864013.1 13 1253
XM_017008525.1 XP_016864014.1 13 1257
XM_017008526.1 XP_016864015.1 9 824
GenBank mRNA AB208943.1 - 15 1175
AF190500.1 - 14 1090
AK122647.1 - 16 1394
AK294694.1 - 14 1198
AK295040.1 - 14 1793
AK300379.1 - 13 1149
AK307889.1 - 13 1192
AK312470.1 - 14 1197
AY899848.1 - 15 1143
AY899849.1 - 12 946
AY899850.1 - 13 991
BC112142.1 - 14 1172 8327610
BC113617.1 - 14 1175 8322669
BC143556.1 - 14 1195
BC143558.1 - 14 1228
BC143559.1 - 13 1097
BX647975.1 - 14 1291
BX647980.1 - 14 1291
BX647985.1 - 14 1320

View Full Product Details