Gene Transcripts
- Gene Symbol
- CDK18
- Entrez Gene ID
- 5129
- Gene Name
- cyclin dependent kinase 18
- Gene Aliases
- PCTAIRE, PCTAIRE3, PCTK3
- Chromosome Location
- Chr. 1: 205504556 - 205532793 on Build GRCh38
- UniGene ID
- Hs.445402
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002596.3 |
NP_002587.2
|
12, 13 |
1530 |
|
NM_212502.2 |
NP_997667.1
|
12, 13 |
1448 |
|
NM_212503.2 |
NP_997668.1
|
12, 13 |
1538 |
|
XM_011509602.2 |
XP_011507904.1
|
12, 13 |
1588 |
|
XM_017001423.1 |
XP_016856912.1
|
12, 13 |
1683 |
|
GenBank mRNA |
AB209307.1 |
- |
10, 11 |
2110 |
|
AK126879.1 |
- |
9 |
1625 |
|
AK293756.1 |
- |
11, 12 |
1812 |
|
AK295272.1 |
- |
10, 11 |
978 |
|
AK296320.1 |
- |
12, 13 |
1496 |
|
AK308913.1 |
- |
10, 11 |
1019 |
|
AL161977.1 |
- |
4, 5 |
745 |
|
AY353237.1 |
- |
12, 13 |
1363 |
|
AY353238.1 |
- |
12, 13 |
1427 |
|
BC000281.2 |
- |
6, 7 |
615 |
|
BC011526.1 |
- |
12, 13 |
1383 |
3855304 |
BC040529.1 |
- |
4, 5 |
841 |
|
BT007299.1 |
- |
11, 12 |
1180 |
|
X66362.1 |
- |
9, 10 |
904 |
|
View Full Product Details