Gene Transcripts
- Gene Symbol
- ZNF330
- Entrez Gene ID
- 27309
- Gene Name
- zinc finger protein 330
- Gene Aliases
- HSA6591, NOA36
- Chromosome Location
- Chr. 4: 141220294 - 141234697 on Build GRCh38
- UniGene ID
- Hs.120766
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ006591.2 |
- |
6 |
516 |
|
View Full Product Details