Gene Transcripts

Gene Symbol
LRRN4CL
Entrez Gene ID
221091
Gene Name
LRRN4 C-terminal like
Gene Aliases
Chromosome Location
Chr. 11: 62686402 - 62689899 on Build GRCh38
UniGene ID
Hs.427449
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203422.3 NP_981967.1 2 1583
GenBank mRNA AK291334.1 - 2 1214
AL834319.1 - 2 1412

View Full Product Details