Gene Transcripts

Gene Symbol
OR10R2
Entrez Gene ID
343406
Gene Name
olfactory receptor family 10 subfamily R member 2
Gene Aliases
OR1-8, OR10R2Q
Chromosome Location
Chr. 1: 158479878 - 158480885 on Build GRCh38
UniGene ID
Hs.451913
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004472.1 NP_001004472.1 1 850

View Full Product Details