Gene Transcripts

Gene Symbol
CCDC171
Entrez Gene ID
203238
Gene Name
coiled-coil domain containing 171
Gene Aliases
C9orf93, bA536D16.1, bA778P13.1
Chromosome Location
Chr. 9: 15552874 - 16062431 on Build GRCh38
UniGene ID
Hs.17267
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173550.2 NP_775821.2 4 584
XM_005251397.2 XP_005251454.1 4 467
XM_005251398.3 XP_005251455.1 4 706
XM_005251399.4 XP_005251456.1 5 766
XM_011517788.2 XP_011516090.1 4 696
XM_011517791.2 XP_011516093.1 4 706
XM_011517796.2 XP_011516098.1 2 113
XM_017014431.1 XP_016869920.1 4 706
XM_017014432.1 XP_016869921.1 4 706
XM_017014433.1 XP_016869922.1 4 706
XM_017014434.1 XP_016869923.1 4 706
XM_017014435.1 XP_016869924.1 4 706
XM_017014436.1 XP_016869925.1 4 706
XM_017014437.1 XP_016869926.1 4 706
XM_017014438.1 XP_016869927.1 4 705
XM_017014439.1 XP_016869928.1 4 706
XM_017014441.1 XP_016869930.1 4 705
XM_017014442.1 XP_016869931.1 4 705
XM_017014443.1 XP_016869932.1 2 108
GenBank mRNA AK308284.1 - 4 467
AY422473.1 - 4 584
BC144214.1 - 3 361 9052734
BC150600.1 - 3 361 9056970
BX537933.1 - 4 445
CR936775.1 - 4 676

View Full Product Details