Gene Transcripts

Gene Symbol
EHD2
Entrez Gene ID
30846
Gene Name
EH domain containing 2
Gene Aliases
PAST2
Chromosome Location
Chr. 19: 47713344 - 47743134 on Build GRCh38
UniGene ID
Hs.744963
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014601.3 NP_055416.2 6 1602
GenBank mRNA AK097126.1 - 6 929
AK097228.1 - 5 921
AK293429.1 - 3 410
AK296907.1 - 7 1494
AK298067.1 - 5 1065
AK300512.1 - 3 553
AK315548.1 - 6 1526
AK316448.1 - 3 502
BC014445.1 - 6 1513 4908085

View Full Product Details