Gene Transcripts

Gene Symbol
ANKRD26P1
Entrez Gene ID
124149
Gene Name
ankyrin repeat domain 26 pseudogene 1
Gene Aliases
Chromosome Location
Chr. 16: 46469337 - 46569097 on Build GRCh38
UniGene ID
Hs.97414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026556.1 10, 11 1616
GenBank mRNA BC070117.1 - 10, 11 1616
CD000265.1 - 2, 3 150
CR749347.1 - 2, 3 112

View Full Product Details