Gene Transcripts
- Gene Symbol
- LRRC52
- Entrez Gene ID
- 440699
- Gene Name
- leucine rich repeat containing 52
- Gene Aliases
- Chromosome Location
- Chr. 1: 165544241 - 165563948 on Build GRCh38
- UniGene ID
- Hs.664975
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098677.1 |
- |
2 |
1017 |
|
View Full Product Details