Gene Transcripts

Gene Symbol
EHBP1
Entrez Gene ID
23301
Gene Name
EH domain binding protein 1
Gene Aliases
HPC12, NACSIN
Chromosome Location
Chr. 2: 62673851 - 63046487 on Build GRCh38
UniGene ID
Hs.271667
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142614.1 NP_001136086.1 10 1582
NM_001142615.2 NP_001136087.1 11 1815
NM_001142616.1 NP_001136088.1 11 1769
NM_015252.3 NP_056067.2 12 1874
XM_005264225.2 XP_005264282.1 12 1772
XM_005264226.2 XP_005264283.1 12 2129
XM_005264227.1 XP_005264284.1 11 1728
XM_005264228.2 XP_005264285.1 12 1833
XM_006711971.2 XP_006712034.1 12 1833
XM_011532713.2 XP_011531015.1 12 1915
XM_011532714.2 XP_011531016.1 12 1764
XM_011532715.2 XP_011531017.1 12 1740
XM_011532716.2 XP_011531018.1 12 1833
XM_011532717.1 XP_011531019.1 12 1833
XM_011532718.2 XP_011531020.1 11 1494
XM_017003643.1 XP_016859132.1 12 1730
XM_017003644.1 XP_016859133.1 12 1707
XM_017003645.1 XP_016859134.1 12 1772
XM_017003646.1 XP_016859135.1 12 1835
XM_017003647.1 XP_016859136.1 11 1667
XM_017003648.1 XP_016859137.1 11 1663
XM_017003649.1 XP_016859138.1 11 1742
XM_017003650.1 XP_016859139.1 12 1772
XM_017003651.1 XP_016859140.1 11 1667
XM_017003652.1 XP_016859141.1 11 1728
XM_017003653.1 XP_016859142.1 11 1656
XM_017003654.1 XP_016859143.1 12 1833
XM_017003655.1 XP_016859144.1 11 1728
GenBank mRNA AB020710.1 - 4 587
AK292545.1 - 11 1815
AL833968.1 - 12 1740
AY331186.1 - 12 1769
AY531390.1 - 10 1287
BC067215.1 - 11 1623 6502257

View Full Product Details