Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017623.4 | NP_060093.3 | 3 | 1478 | |
NM_199078.2 | NP_951060.1 | 2 | 1334 | ||
XM_011510957.2 | XP_011509259.1 | 3 | 1838 | ||
XM_017003800.1 | XP_016859289.1 | 3 | 1838 | ||
GenBank mRNA | AF216965.1 | - | 3 | 577 | |
AK126847.1 | - | 3 | 1478 | ||
BC007199.1 | - | 2 | 490 | 2964706 | |
BC018604.1 | - | 2 | 223 | ||
BC037272.1 | - | 3 | 1487 | 4823510 | |
CR749702.1 | - | 2 | 112 |