Gene Transcripts

Gene Symbol
Tm7sf3
Entrez Gene ID
297725
Gene Name
transmembrane 7 superfamily member 3
Gene Aliases
Chromosome Location
Chr. 4: 180913703 - 180951564 on Build Rnor_6.0
UniGene ID
Rn.23905
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011970.1 NP_001011970.1 12 1938
GenBank mRNA BC090030.1 - 12 1938

View Full Product Details