Gene Transcripts
- Gene Symbol
- KRTAP5-3
- Entrez Gene ID
- 387266
- Gene Name
- keratin associated protein 5-3
- Gene Aliases
- KRTAP5-9, KRTAP5.3
- Chromosome Location
- Chr. 11: 1607565 - 1608463 on Build GRCh38
- UniGene ID
- Hs.532471
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB126072.1 |
- |
1 |
16 |
|
View Full Product Details