Gene Transcripts

Gene Symbol
Tbc1d22b
Entrez Gene ID
502414
Gene Name
TBC1 domain family, member 22B
Gene Aliases
Chromosome Location
Chr. 20: 8216097 - 8278344 on Build Rnor_6.0
UniGene ID
Rn.16811
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025059.1 NP_001020230.1 4 627
GenBank mRNA AY380821.1 - 4 627

View Full Product Details