Gene Transcripts

Gene Symbol
Cnksr3
Entrez Gene ID
308113
Gene Name
Cnksr family member 3
Gene Aliases
Magi1, Prp4
Chromosome Location
Chr. 1: 43789797 - 43884267 on Build Rnor_6.0
UniGene ID
Rn.137681
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012061.1 NP_001012061.1 11 1607
XM_006227875.2 XP_006227937.1 11 2764
GenBank mRNA AY328890.1 - 11 1607
AY333962.1 - 3 237

View Full Product Details