Gene Transcripts

Gene Symbol
Lrrc52
Entrez Gene ID
240899
Gene Name
leucine rich repeat containing 52
Gene Aliases
4930413P14Rik
Chromosome Location
Chr. 1: 167445675 - 167466780 on Build GRCm38
UniGene ID
Mm.159799
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013382.2 NP_001013400.1 1 553
GenBank mRNA BC087947.1 - 1 553

View Full Product Details