Gene Transcripts
- Gene Symbol
- GOLGA6L9
- Entrez Gene ID
- 440295
- Gene Name
- golgin A6 family-like 9
- Gene Aliases
- GOLGA6L20
- Chromosome Location
- Chr. 15: 82429363 - 82439387 on Build GRCh38
- UniGene ID
- Hs.498322
- Species
- Homo sapiens
- Gene Symbol
- GOLGA6L4
- Entrez Gene ID
- 643707
- Gene Name
- golgin A6 family-like 4
- Gene Aliases
- Chromosome Location
- Chr. 15: 84235773 - 84245368 on Build GRCh38
- UniGene ID
- Hs.498345
- Species
- Homo sapiens
- Gene Symbol
- GOLGA6L5P
- Entrez Gene ID
- 374650
- Gene Name
- golgin A6 family-like 5, pseudogene
- Gene Aliases
- GOLGA6L5, GOLGA6L8
- Chromosome Location
- Chr. 15: 84506168 - 84516847 on Build GRCh38
- UniGene ID
- Hs.454625
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003246.4 |
|
10 |
3964 |
|
GenBank mRNA |
BX647899.1 |
- |
1 |
2365 |
|
View Full Product Details