Gene Transcripts

Gene Symbol
IMPACT
Entrez Gene ID
55364
Gene Name
impact RWD domain protein
Gene Aliases
RWDD5
Chromosome Location
Chr. 18: 24426645 - 24453531 on Build GRCh38
UniGene ID
Hs.515317
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018439.3 NP_060909.1 11 2483
XM_017025832.1 XP_016881321.1 11 2736
GenBank mRNA AB026264.1 - 11 2410

View Full Product Details