Gene Transcripts
- Gene Symbol
- BRK1
- Entrez Gene ID
- 55845
- Gene Name
- BRICK1, SCAR/WAVE actin nucleating complex subunit
- Gene Aliases
- C3orf10, HSPC300, MDS027, hHBrk1
- Chromosome Location
- Chr. 3: 10115649 - 10127190 on Build GRCh38
- UniGene ID
- Hs.649307
- Species
- Homo sapiens
- Gene Symbol
- LOC100130542
- Entrez Gene ID
- 100130542
- Gene Name
- uncharacterized LOC100130542
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.678537
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127083.1 |
- |
|
4322 |
|
View Full Product Details