Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001045477.2 | NP_001038942.1 | 2 | 636 | |
NM_001170741.1 | NP_001164212.1 | 2 | 636 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017561.1 | NP_060031.1 | 2 | 636 | |
GenBank mRNA | AL133071.1 | - | 1 | 189 | |
BC130390.1 | - | 2 | 636 | 40146421 | |
BC143798.1 | - | 2 | 636 | 9052312 |