Gene Transcripts

Gene Symbol
MAP7D3
Entrez Gene ID
79649
Gene Name
MAP7 domain containing 3
Gene Aliases
MDP3
Chromosome Location
Chr. X: 136213220 - 136256482 on Build GRCh38
UniGene ID
Hs.446275
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173517.1 NP_001166988.1 18 3204
GenBank mRNA AL832120.1 - 18 2996

View Full Product Details